Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APE Antibody (CPTC-APEX1-2), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB381069 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB381069 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB381069 Supplier Novus Biologicals Supplier No. NBP308370JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

APE Monoclonal antibody specifically detects APE in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen APE
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone CPTC-APEX1-2
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias AP endonuclease 1, APE, APE1, APE-1, APEAPEX nuclease (multifunctional DNA repair enzyme), APENAP endonuclease class I, APEX deoxyribonuclease (apurinic or apyrimidinic), APEX nuclease, APEX nuclease (multifunctional DNA repair enzyme) 1, Apurinic-apyrimidinic endonuclease 1, APXAP lyase, DNA-(apurinic or apyrimidinic site) lyase, EC 3.1, EC 4.2.99.18, HAP1apurinic/apyrimidinic (abasic) endonuclease, multifunctional DNA repair enzyme, protein REF-1, redox factor 1, Redox factor-1, REF-1, REF1 apurinic/apyrimidinic exonuclease
Host Species Mouse
Immunogen Recombinant human full-length APE protein (Uniprot: P27695)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Autophagy, Base Excision Repair, Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 328
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.