Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APE Mouse anti-Human, Clone: CPTC-APEX1-2, Novus Biologicals™
SDP

Catalog No. NB123202 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB123202 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB123202 Supplier Novus Biologicals Supplier No. NBP30786420UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

APE Monoclonal specifically detects APE in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen APE
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone CPTC-APEX1-2
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Alias AP endonuclease 1, APE, APE1, APE-1, APEAPEX nuclease (multifunctional DNA repair enzyme), APENAP endonuclease class I, APEX deoxyribonuclease (apurinic or apyrimidinic), APEX nuclease, APEX nuclease (multifunctional DNA repair enzyme) 1, Apurinic-apyrimidinic endonuclease 1, APXAP lyase, DNA-(apurinic or apyrimidinic site) lyase, EC 3.1, EC 4.2.99.18, HAP1apurinic/apyrimidinic (abasic) endonuclease, multifunctional DNA repair enzyme, protein REF-1, redox factor 1, Redox factor-1, REF-1, REF1 apurinic/apyrimidinic exonuclease
Host Species Mouse
Immunogen Recombinant human full-length APE protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Autophagy, Base Excision Repair, Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 328
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.