Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APEG1 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB090629 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB090629 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB090629 Supplier Novus Biologicals Supplier No. NBP299997UV

Rabbit Polyclonal Antibody

APEG1 Polyclonal specifically detects APEG1 in Human samples. It is validated for ELISA.

Specifications

Antigen APEG1
Applications ELISA
Classification Polyclonal
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias aortic preferentially expressed gene 1, Aortic preferentially expressed protein 1, APEG1, BPEG, EC 2.7.11, EC 2.7.11.1, KIAA1297APEG-1, MGC12676, nuclear protein, marker for differentiated aortic smooth muscle anddown-regulated with vascular injury, SPEG complex locus, SPEGalpha, SPEGbeta, striated muscle preferentially expressed protein kinase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant APEG1 (Q15772-4; Met1-Glu113).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10290
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.