Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APH1A Antibody, Novus Biologicals™
SDP

Catalog No. NBP227176SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227176SS 0.025 mg
NBP227176 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227176SS Supplier Novus Biologicals Supplier No. NBP227176SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

APH1A Polyclonal specifically detects APH1A in Human, Bovine, Canine, Equine, Opossum, Primate samples. It is validated for Western Blot.

Specifications

Antigen APH1A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 4-6ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias 6530402N02Rik, anterior pharynx defective 1 homolog A (C. elegans), APH-1, APH-1A, aph-1alpha, CGI-78, gamma-secretase subunit APH-1A, Presenilin-stabilization factor, PSF
Gene Symbols APH1A
Host Species Rabbit
Immunogen A portion of amino acids 80-145 of human APH1A was used as the immunogen.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline GPCR, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 51107
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, monkey, mouse, cow and dog, 94% homologous in rat and 88% homologous in chicken.
Target Species Human, Bovine, Canine, Equine, Opossum, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.