Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APH1A Antibody, Novus Biologicals™
SDP

Catalog No. NB10074360 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074360 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074360 Supplier Novus Biologicals Supplier No. NB10074360
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

APH1A Polyclonal specifically detects APH1A in Human, Mouse samples. It is validated for ChIP Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen APH1A
Applications ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Formulation PBS, 1.0 mg/ml BSA and 30% Glycerol with 0.005% Sodium Azide
Gene Accession No. Q96BI3
Gene Alias 6530402N02Rik, anterior pharynx defective 1 homolog A (C. elegans), APH-1, APH-1A, aph-1alpha, CGI-78, gamma-secretase subunit APH-1A, Presenilin-stabilization factor, PSF
Gene Symbols APH1A
Host Species Rabbit
Immunogen Synthetic peptide, KLH conjugated, corresponding to residues C (245) R R Q E D S R V M V Y S A L R I P P E D (265) of human APH-1aL/CT.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline GPCR, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 51107
Test Specificity APH-1
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.