Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APIP Antibody (19F461), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB10056567Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056567Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056567Z Supplier Novus Biologicals Supplier No. NB10056567AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

APIP Monoclonal specifically detects APIP in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen APIP
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 19F461
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q96GX9
Gene Alias APAF1 interacting protein, APAF1-interacting protein, APIP2, CGI29, CGI-29, dJ179L10.2, EC 4.2.1.109, MMRP19, MTRu-1-P dehydratase, probable methylthioribulose-1-phosphate dehydratase
Gene Symbols APIP
Host Species Mouse
Immunogen The antibody was developed against a full-length His-tagged recombinant APIP.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 51074
Test Specificity This monoclonal antibody can be used for detection of APIP and APIP2.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.