Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APJ/Apelin receptor Antibody (72133R) [DyLight 680], Novus Biologicals™
SDP

Catalog No. NB372901 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372901 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372901 Supplier Novus Biologicals Supplier No. FAB8561Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

APJ/Apelin receptor Monoclonal specifically detects APJ/Apelin receptor in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.

Specifications

Antigen APJ/Apelin receptor
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 72133R
Conjugate DyLight 680
Dilution Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias AGTRL1HG11, angiotensin II receptor-like 1, Angiotensin receptor-like 1, apelin receptor, APJ (apelin) receptor, APJ receptor, APJFLJ96609, APJR, FLJ90771, G protein-coupled receptor APJ, G-protein coupled receptor APJ, G-protein coupled receptor HG11, HG11 orphan receptor, MGC45246
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human APJ, Met1-Asp380, Accession # P35414
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 187
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.