Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APLF Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | APLF |
---|---|
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
APLF Polyclonal specifically detects APLF in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
APLF | |
Polyclonal | |
Rabbit | |
Human | |
Q8IW19 | |
200558 | |
This antibody was developed against a recombinant protein corresponding to amino acids: SQLEGSTEIAKTQMTPTNSVSFLGENRDCNKQQPILAERKRILPTWMLAEHLSDQNLSVPAISGGNVIQGSGKEEICKDKSQLN | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
APFL, aprataxin- and PNK-like factor, aprataxin and PNK-like factor, aprataxin and PNKP like factor, Apurinic-apyrimidinic endonuclease APLF, C2orf13FLJ16593, chromosome 2 open reading frame 13, EC 4.2.99.18, MGC47799, PALF, PNK and APTX-like FHA domain-containing protein, PNK and APTX-like FHA protein, Xip1, XRCC1-interacting protein 1 | |
APLF | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title