Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APLP-1 Antibody (354008), PE, Novus Biologicals™
SDP

Catalog No. NB363221 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363221 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363221 Supplier Novus Biologicals Supplier No. FAB3129P
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

APLP-1 Monoclonal antibody specifically detects APLP-1 in Human samples. It is validated for Immunocytochemistry

Specifications

Antigen APLP-1
Applications Immunocytochemistry
Classification Monoclonal
Clone 354008
Conjugate PE
Dilution Immunocytochemistry
Formulation PBS
Gene Alias amyloid beta (A4) precursor-like protein 1, amyloid precursor-like protein 1, amyloid-like protein 1, APLPAPLP-1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human APLP-1, Gln34-Glu580 (predicted), Accession # P51693
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 333
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.