Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APLP-2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB319783 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB319783 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB319783 Supplier Novus Biologicals Supplier No. NBP298659MFV500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

APLP-2 Polyclonal antibody specifically detects APLP-2 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen APLP-2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias amyloid beta (A4) precursor-like protein 2, amyloid precursor protein homolog HSD-2, Amyloid protein homolog, amyloid-like protein 2, APLP-2, APPHCDEI box-binding protein, APPL2, CDEBP
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human APLP-2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 334
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.