Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APM2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB151786 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151786 25 μg
NB151785 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB151786 Supplier Novus Biologicals Supplier No. NBP31191625UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

APM2 Polyclonal antibody specifically detects APM2 in Human samples. It is validated for Western Blot

Specifications

Antigen APM2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 - 5 ug/ml
Formulation PBS
Gene Alias adipose specific 2, apM-2, APM2, chromosome 10 open reading frame 116
Host Species Rabbit
Immunogen Rabbits were immunized with a synthetic peptide derived from human APM2 protein (Uniprot: Q15847)
Purification Method Ammonium sulfate precipitation
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10974
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.