Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APM2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$231.50 - $499.50
Specifications
Antigen | APM2 |
---|---|
Dilution | Western Blot 1 - 5 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
APM2 Polyclonal antibody specifically detects APM2 in Human samples. It is validated for Western BlotSpecifications
APM2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
adipose specific 2, apM-2, APM2, chromosome 10 open reading frame 116 | |
Rabbits were immunized with a synthetic peptide derived from human APM2 protein (Uniprot: Q15847) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot 1 - 5 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS | |
10974 | |
IgG | |
Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title