Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Apo-BrdU Apoptosis Detection Kit

Catalog No. 509273
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-927-3 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 50-927-3 Supplier Invitrogen™ Supplier No. 88667188
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains Sodium azide

Contains the instructions and reagents required for measuring apoptosis in cells. APO-BRDU™ Apoptosis Detection Kit is a 2-color staining method for labeling DNA breaks and total cellular DNA to detect apoptotic cells by flow cytometric analysis in immunochemistry applications.

The APO-BRDU™ Apoptosis Detection Kit includes sufficient reagents to process a total of 60 cell suspensions, including:

  • 5mL positive and 5 mL negative control cell suspensions of approximately 1×106 cells per mL in 70% (v/v) ethanol for assessing reagent performance
  • Washing, reaction and rinsing buffers for processing individual steps in the assay
  • Terminal deoxynucleotidyl transferase enzyme (TdT), bromodeoxyuridine triphosphate (Br-dUTP), and fluorescein labeled anti-BrdU antibody for labeling DNA breaks
  • Propidium iodide/RNase A solution for counterstaining the total DNA

One of the most easily measured features of apoptotic cells is the break-up of the genomic DNA by cellular nucleases. These DNA fragments can be extracted from apoptotic cells and result in the appearance of DNA laddering when the DNA is analyzed by agarose gel electrophoresis. The DNA of non-apoptotic cells that remains largely intact does not display this laddering on agarose gels during electrophoresis.

The large number of DNA fragments appearing in apoptotic cells results in a multitude of 3′-hydroxyl termini in the DNA. This property can be used to identify apoptotic cells by labeling the 3′-hydroxyl ends with bromolated deoxyuridine triphosphate nucleotides (Br-dUTP). The enzyme terminal deoxynucleotidyl transferase (TdT) catalyzes a template independent addition of deoxyribonucleoside triphosphates to the 3′-hydroxyl ends of double- or single-stranded DNA with either blunt, recessed or overhanging ends. A substantial number of these sites are available in apoptotic cells providing the basis for the method utilized in the APO-BRDU™ Kit.

Recent evidence has demonstrated that Br-dUTP is more readily incorporated into the genome of apoptotic cells than are the deoxynucleotide triphosphates complexed to larger ligands like fluorescein, biotin or digoxigenin. This greater incorporation gives rise to a brighter flow cytometry signal when the Br-dUTP sites are identified by a fluorescein labeled anti-BrdU monoclonal antibody. Non-apoptotic cells do not incorporate significant amounts of the Br-dUTP due to the lack of exposed 3′-hydroxyl DNA ends.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 kit
Format Kit
Product Type Apoptosis Detection Kit
For Use With (Application) Flow Cytometry
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.