Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOA1BP Antibody, Novus Biologicals™
SDP

Catalog No. NBP120877 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP120877 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP120877 Supplier Novus Biologicals Supplier No. NBP120877
Add to Cart
Add to Cart

Goat Polyclonal Antibody

APOA1BP Polyclonal specifically detects APOA1BP in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen APOA1BP
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1.5 ug/ml, Peptide ELISA Detection limit 1:1000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q8NCW5
Gene Alias AIBPAI-BP, apolipoprotein A-I binding protein, apolipoprotein A-I-binding protein, MGC119143, MGC119144, MGC119145, YjeF N-terminal domain-containing protein 1, YjeF_N1, YJEFN1apoA-I binding protein
Gene Symbols NAXE
Host Species Goat
Immunogen Peptide with sequence PPYPDTECVYRLQ corresponding to C-Terminus according to NP_658985.2.
Molecular Weight of Antigen 32 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128240
Test Specificity Human. Expected cross-reactivity from sequence similarity: Mouse, Rat, Dog, Cow and Pig.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.