Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOB48R Antibody (OTI1B1), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N272052A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272052A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272052A700 Supplier Novus Biologicals Supplier No. NBP272052AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

APOB48R Monoclonal specifically detects APOB48R in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen APOB48R
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1B1
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias APOB100R, apoB-48R, APOB48Rapolipoprotein B48 receptor, apolipoprotein B receptor, Apolipoprotein B-100 receptor, Apolipoprotein B-48 receptor
Gene Symbols APOBR
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 780-1088 of human APOB48R (NP_061160) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 55911.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.