Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOBEC3B Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017384 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017384 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017384 Supplier Novus Biologicals Supplier No. NBP282073
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

APOBEC3B Polyclonal specifically detects APOBEC3B in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen APOBEC3B
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias APOBEC1L, apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3B, ARCD3, ARP4, bK150C2.2, cytidine deaminase, DJ742C19.2, EC 3.5.4, EC 3.5.4.-, FLJ21201, phorbolin 3, Phorbolin-1-related protein, phorbolin-2/3, PHRBNLphorbolin 2, probable DNA dC->dU-editing enzyme APOBEC-3B
Gene Symbols APOBEC3B
Host Species Rabbit
Immunogen APOBEC3B antibody was raised against a 14 amino acid peptide near the amino terminus of human APOBEC3B. The immunogen is located within amino acids 20 - 70 of APOBEC3B. Amino Acid Squence: EVKIKRGRSNLLWD
Molecular Weight of Antigen 48 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9582
Test Specificity APOBEC3B antibody is human, mouse and rat reactive . At least three isoforms of APOBEC3B are known to exist; this antibody will detect all three. APOBEC3B antibody may cross-react with APOBEC3D.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.