Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APOD Mouse anti-Human, Clone: 1C6D10, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 662151IG150UL

Description
Apolipoprotein D (APOD) is a component of high density lipoprotein that has no marked similarity to other apolipoprotein sequences. It has a high degree of homology to plasma retinol-binding protein and other members of the alpha 2 microglobulin protein superfamily of carrier proteins, also known as lipocalins. This glycoprotein is closely associated with the enzyme lecithin:cholesterol acyltransferase - an enzyme involved in lipoprotein metabolism.Specifications
APOD | |
Monoclonal | |
0.94 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P05090 | |
APOD | |
APOD Fusion Protein Ag21422 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1C6D10 | |
Unconjugated | |
APOD | |
Apo D, APOD, apolipoprotein D | |
Mouse | |
Protein G | |
RUO | |
347 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction