Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APOD Mouse anti-Human, Clone: 1C6D10, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | APOD |
---|---|
Clone | 1C6D10 |
Concentration | 0.94 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6648
![]() ![]() |
Proteintech Group Inc
662151IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Apolipoprotein D (APOD) is a component of high density lipoprotein that has no marked similarity to other apolipoprotein sequences. It has a high degree of homology to plasma retinol-binding protein and other members of the alpha 2 microglobulin protein superfamily of carrier proteins, also known as lipocalins. This glycoprotein is closely associated with the enzyme lecithin:cholesterol acyltransferase - an enzyme involved in lipoprotein metabolism.Specifications
APOD | |
0.94 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Apo D, APOD, apolipoprotein D | |
APOD | |
IgG1 | |
Protein G | |
Antibody |
1C6D10 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P05090 | |
347 | |
APOD Fusion Protein Ag21422 | |
Primary | |
-20°C | |
APOD |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title