Learn More
Invitrogen™ APOE Monoclonal Antibody (GT1627)

Description
This product can be used in Sandwich ELISA to detect APOE using MA5-50301 as a capture antibody and PA5-27088 as the detection antibody.
Specifications
Specifications
| Antigen | APOE |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | GT1627 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and no preservative |
| Gene | APOE |
| Gene Accession No. | P02649, P02650 |
| Gene Alias | AD2; AI255918; APOE; apo-E; ApoE4; APOEA; Apolipoprotein; apolipoprotein E; apolipoprotein E3; apolipoprotein E4; Apoprotein; B2G1; BG; LDLCQ5; LPG; MGC1571 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.