Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ApoE4 Antibody (4E4), HRP, Novus Biologicals™
SDP

Catalog No. NBP149529H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149529H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149529H Supplier Novus Biologicals Supplier No. NBP149529H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ApoE4 Monoclonal specifically detects ApoE4 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen ApoE4
Applications Western Blot, ELISA
Classification Monoclonal
Clone 40000
Conjugate HRP
Dilution Western Blot, ELISA
Gene Accession No. P02649
Gene Alias Apo-E, apolipoprotein E, apolipoprotein E3, late onset), MGC1571
Gene Symbols APOE
Host Species Mouse
Immunogen Synthetic peptide made to an internal region of human ApoE4 (within residues 100-150) [Uniprot: P02649]
Molecular Weight of Antigen 34 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 348
Test Specificity This antibody is specific for ApoE4. It does not cross-react with the ApoE2 or ApoE3 isoforms.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.