Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ApoE4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP150669 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP150669 0.1 mg
NB018659 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP150669 Supplier Novus Biologicals Supplier No. NBP150669

Rabbit Polyclonal Antibody

ApoE4 Polyclonal specifically detects ApoE4 in Human samples. It is validated for Western Blot.

Specifications

Antigen ApoE4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS (pH 7.2), 0.5% BSA, 30% glycerol with 0.01% Thimerosal
Gene Accession No. P02649
Gene Alias Apo-E, apolipoprotein E, apolipoprotein E3, late onset), MGC1571
Gene Symbols APOE
Host Species Rabbit
Immunogen Recombinant human ApoE4 (AAB59397).
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.