Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ APOL2 Monoclonal Antibody (C1)
Mouse Monoclonal Antibody
$243.00 - $552.50
Specifications
Antigen | APOL2 |
---|---|
Clone | C1 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541555
|
Invitrogen™
MA541555 |
200 μL |
Each for $552.50
|
|
|||||
PIMA543956
|
Invitrogen™
MA543956 |
20 μL |
Each for $243.00
|
|
|||||
Description
APOL2 Monoclonal Antibody for Western Blot, IHC (P)
This gene is a member of the apolipoprotein L gene family. The encoded protein is found in the cytoplasm, where it may affect the movement of lipids or allow the binding of lipids to organelles. Two transcript variants encoding the same protein have been found for this gene.Specifications
APOL2 | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9BQE5 | |
23780 | |
Recombinant protein Apolipoprotein L2. The antigen corresponds to amino acid range 186-337 of the target protein. | |
Antibody |
C1 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG1 κ | |
Human | |
APOL2 | |
Apol2; ApoL-2; APOL3; Apol8; APOL-II; apolipoprotein L 8; apolipoprotein L, 2; Apolipoprotein L2; Apolipoprotein L-2; apolipoprotein L-II; OTTHUMP00000028936 | |
APOL2 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title