Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APOLD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | APOLD1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17933020
![]() |
Novus Biologicals
NBP17933020UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179330
![]() |
Novus Biologicals
NBP179330 |
100 μL |
Each for $487.50
|
|
|||||
Description
APOLD1 Polyclonal specifically detects APOLD1 in Human samples. It is validated for Western Blot.Specifications
APOLD1 | |
Polyclonal | |
Rabbit | |
Q96LR9 | |
81575 | |
Synthetic peptide directed towards the middle region of human APOLD1The immunogen for this antibody is APOLD1. Peptide sequence LIPRRAEGDTKVSQAVLKAKIQKLAESLESCTGALDELSEQLESRVQLCT. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
apolipoprotein L domain containing 1, apolipoprotein L domain-containing protein 1, DKFZP434F0318, FLJ25138, FLJ95166, Vascular early response gene protein, VERGEDKFZp434F0318 | |
APOLD1 | |
IgG | |
30 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title