Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein A-I/ApoA1 Antibody (031), PE, Novus Biologicals™
SDP

Catalog No. NB306894 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306894 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB306894 Supplier Novus Biologicals Supplier No. NBP289599PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Apolipoprotein A-I/ApoA1 Monoclonal antibody specifically detects Apolipoprotein A-I/ApoA1 in Human samples. It is validated for ELISA

Specifications

Antigen Apolipoprotein A-I/ApoA1
Applications ELISA
Classification Monoclonal
Clone 031
Conjugate PE
Dilution Sandwich ELISA Capture
Formulation PBS
Gene Alias APOA1, Apo-AI, ApoA-I, Apolipoprotein A1, apolipoprotein A-I, MGC117399
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Apolipoprotein A-I/ApoA1 (Uniprot#: P02647; Asp25-Gln267).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Breast Cancer, Cancer, Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 335
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.