Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein A-I/ApoA1 Antibody (1E12), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP197943AG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197943AG 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197943AG Supplier Novus Biologicals Supplier No. NBP197943AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Apolipoprotein A-I/ApoA1 Monoclonal specifically detects Apolipoprotein A-I/ApoA1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Apolipoprotein A-I/ApoA1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1000000000000
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias APOA1, apoA-I, Apo-AI, Apolipoprotein A1, apolipoprotein A-I, MGC117399
Gene Symbols APOA1
Host Species Mouse
Immunogen Recombinant human Apolipoprotein A1 (25-267aa) purified from E. coli
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Breast Cancer, Cancer, Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 335
Test Specificity Anti-human Apolipoprotein A1 mAb, clone 1E12, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human APOA1 protein.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.