Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein A-I/ApoA1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP215429 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215429 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215429 Supplier Novus Biologicals Supplier No. NBP215429
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Apolipoprotein A-I/ApoA1 Polyclonal antibody specifically detects Apolipoprotein A-I/ApoA1 in Human, Mouse, Rat, Golden Syrian Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).

Specifications

Antigen Apolipoprotein A-I/ApoA1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:20000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS pH7, 20% Glycerol with 0.025% Proclin 300
Gene Accession No. P02647
Gene Alias APOA1, apoA-I, Apo-AI, Apolipoprotein A1, apolipoprotein A-I, MGC117399
Gene Symbols APOA1
Host Species Rabbit
Immunogen Full length human Apolipoprotein A1 Recombinant protein.
Molecular Weight of Antigen 31 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 335
Target Species Human, Mouse, Rat, Golden Syrian Hamster
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.