Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein A-II/ApoA2 Antibody, Novus Biologicals™
SDP

Catalog No. NB6001303 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001303 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001303 Supplier Novus Biologicals Supplier No. NB6001303

Goat Polyclonal Antibody

Apolipoprotein A-II/ApoA2 Polyclonal specifically detects Apolipoprotein A-II/ApoA2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Apolipoprotein A-II/ApoA2
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:10000, ELISA 1:5000-1:10000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:10-1:500
Formulation 0.125 M Sodium Borate, 0.075 M Sodium Chloride, 0.005 M EDTA, pH 8.0 with 0.01% Sodium Azide
Gene Accession No. P02652, P02652, P02652, P02652
Gene Alias APOA2, apoAII, Apo-AII, ApoA-II, Apolipoprotein A2, apolipoprotein A-II
Gene Symbols APOA2
Host Species Goat
Immunogen Apolipoprotein A-II/ApoA2 was isolated from human plasma by density gradient centrifugation followed by HPLC purification. (Uniprot: P02652)
Molecular Weight of Antigen 11.17 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 336
Test Specificity Typically less than 1% cross reactivity against other types of apolipoprotein was detected by ELISA against purified standards. This antibody reacts with human apolipoprotein A-II and has negligible cross-reactivity with Type A-I, B, C-I, C-II, C-III, E and J apolipoproteins. Specific cross reaction of anti-apolipoprotein antibodies with antigens from other species has not been determined. Non-specific cross reaction of anti-apolipoprotein antibodies with other human serum proteins is negligible.
Target Species Human
Content And Storage Store at 4°C short term. For extended storage, add an equal volume of glycerol, aliquot and store at -20°C or below. Avoid repeated freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.