Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein A-II/ApoA2 Rabbit anti-Human, Mouse, Rat, Clone: 5F7U1, Novus Biologicals™
SDP

Catalog No. NB167428 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167428 20 μg
NB167427 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167428 Supplier Novus Biologicals Supplier No. NBP31616820UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Apolipoprotein A-II/ApoA2 Monoclonal antibody specifically detects Apolipoprotein A-II/ApoA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence

Specifications

Antigen Apolipoprotein A-II/ApoA2
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 5F7U1
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias APOA2, apoAII, Apo-AII, ApoA-II, Apolipoprotein A2, apolipoprotein A-II
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human Apolipoprotein A-II/ApoA2 (P02652).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 336
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.