Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein A-IV/ApoA4 Antibody (APOA4/3372), FITC, Novus Biologicals™
SDP

Catalog No. NB389140 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389140 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389140 Supplier Novus Biologicals Supplier No. NBP308947F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Apolipoprotein A-IV/ApoA4 Monoclonal antibody specifically detects Apolipoprotein A-IV/ApoA4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen Apolipoprotein A-IV/ApoA4
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone APOA4/3372
Conjugate FITC
Formulation PBS
Gene Alias apo-AIV, ApoA-IV, Apolipoprotein A4, apolipoprotein A-IV, MGC142154, MGC142156
Host Species Mouse
Immunogen Human recombinant Apolipoprotein A-IV/ApoA4 protein fragment (around aa 239-371) (exact sequence is proprietary) (Uniprot: P06727)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 337
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.