Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein A5 Antibody (4H8H8E2), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. N1107307A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1107307A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1107307A94 Supplier Novus Biologicals Supplier No. NB11057307AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Apolipoprotein A5 Monoclonal specifically detects Apolipoprotein A5 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Apolipoprotein A5
Applications Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 4H8H8E2
Conjugate Alexa Fluor 594
Dilution Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Apo-AV, ApoA-V, APOAV, Apolipoprotein A5, apolipoprotein A-V, FLJ97995, MGC126836, RAP3MGC126838, Regeneration-associated protein 3
Gene Symbols APOA5
Host Species Mouse
Immunogen Ni-NTA purified full-length recombinant human Apolipoprotein A5 expressed in E. Coli strain BL21 (DE3). [UniProtNo. Q6Q788]
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 116519.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.