Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein A5 Antibody (4H8H8E2), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 1 publication

Supplier:  Novus Biologicals NB400138SS

 View more versions of this product

Catalog No. NB400138SS


Only null left
Add to Cart

Description

Description

Apolipoprotein A5 Monoclonal specifically detects Apolipoprotein A5 in Human samples. It is validated for ELISA.
Specifications

Specifications

Apolipoprotein A5
Monoclonal
Unconjugated
Ascites with 0.1% Sodium Azide
Apo-AV, ApoA-V, APOAV, Apolipoprotein A5, apolipoprotein A-V, FLJ97995, MGC126836, RAP3MGC126838, Regeneration-associated protein 3
Mouse
Unpurified
RUO
Primary
Human. Immunogen sequence has 83% identity to Bovine, 79% identity to rat and 78% identity to mouse.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG1 κ
ELISA
4H8H8E2
ELISA 1:100-1:2000
Q6Q788
APOA5
Ni-NTA purified full-length recombinant human Apolipoprotein A5 expressed in E. Coli strain BL21 (DE3). [UniProt# Q6Q788]
0.025 mL
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
116519
Human
Ascites
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only