Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein A5 Antibody (4H8H8E2), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$181.90 - $366.00
Specifications
| Antigen | Apolipoprotein A5 |
|---|---|
| Clone | 4H8H8E2 |
| Dilution | ELISA 1:100-1:2000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB400138SS
![]() |
Novus Biologicals
NB400138SS |
0.025 mL |
Each for $181.90
|
|
|||||
NB400138
![]() |
Novus Biologicals
NB400138 |
0.1 mL |
Each for $366.00
|
|
|||||
Description
Apolipoprotein A5 Monoclonal specifically detects Apolipoprotein A5 in Human samples. It is validated for ELISA.Specifications
| Apolipoprotein A5 | |
| ELISA 1:100-1:2000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| Q6Q788 | |
| 116519 | |
| Ni-NTA purified full-length recombinant human Apolipoprotein A5 expressed in E. Coli strain BL21 (DE3). [UniProt# Q6Q788] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| 4H8H8E2 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| Ascites with 0.1% Sodium Azide | |
| Apo-AV, ApoA-V, APOAV, Apolipoprotein A5, apolipoprotein A-V, FLJ97995, MGC126836, RAP3MGC126838, Regeneration-associated protein 3 | |
| APOA5 | |
| IgG1 κ | |
| Unpurified | |
| Human. Immunogen sequence has 83% identity to Bovine, 79% identity to rat and 78% identity to mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title