Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein B/ApoB Antibody (APOB/4333), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB392666 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392666 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392666 Supplier Novus Biologicals Supplier No. NBP308230JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Apolipoprotein B/ApoB Monoclonal antibody specifically detects Apolipoprotein B/ApoB in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen Apolipoprotein B/ApoB
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone APOB/4333
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Apo B-100, apoB-100, apoB-48, apolipoprotein B (including Ag(x) antigen), apolipoprotein B-100, apolipoprotein B48, FLDB, LDLCQ4, mutant Apo B 100
Host Species Mouse
Immunogen Human recombinant Apolipoprotein B/ApoB protein fragment (around aa 592-689) (exact sequence is proprietary) (Uniprot: P04114)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 338
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.