Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein B/ApoB Antibody, Novus Biologicals™
SDP

Catalog No. NB200527 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200527 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB200527 Supplier Novus Biologicals Supplier No. NB200527
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

Apolipoprotein B/ApoB Polyclonal specifically detects Apolipoprotein B/ApoB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Apolipoprotein B/ApoB
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunoprecipitation
Formulation Whole antisera with 0.02% Sodium Azide
Gene Alias Apo B-100, apoB-100, apoB-48, apolipoprotein B (including Ag(x) antigen), apolipoprotein B-100, apolipoprotein B48, FLDB, LDLCQ4, mutant Apo B 100
Gene Symbols APOB
Host Species Rabbit
Immunogen Mouse low density lipoprotein (LDL)
Purification Method Unpurified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 338
Test Specificity Recognizes mouse as well as human Apo B48 and B100 as these proteins are highly conserved.
Target Species Human, Mouse
Content And Storage Store at -70C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.