Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein D Antibody (APOD/3413), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB179447 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB179447 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB179447 Supplier Novus Biologicals Supplier No. NBP314089JF549
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Apolipoprotein D Monoclonal antibody specifically detects Apolipoprotein D in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Apolipoprotein D
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone APOD/3413
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias apo-D, ApoD, apolipoprotein D
Host Species Mouse
Immunogen Human recombinant Apolipoprotein D protein fragment (around aa21-119) (exact sequence is proprietary) (Uniprot: P05090)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 347
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.