Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein E/ApoE Mouse anti-Non-human Primate, Clone: E981, Novus Biologicals™
SDP

Catalog No. NB171935 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1000 μg
250 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171935 1000 μg
NB171936 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB171935 Supplier Novus Biologicals Supplier No. NBP3182971000UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Apolipoprotein E/ApoE Monoclonal antibody specifically detects Apolipoprotein E/ApoE in Non-human Primate samples. It is validated for Western Blot, ELISA

Specifications

Antigen Apolipoprotein E/ApoE
Applications Western Blot, ELISA
Classification Monoclonal
Clone E981
Concentration 0.5 mg/ml
Conjugate Unconjugated
Formulation 0.2 um filtered PBS
Gene Alias Apo-E, apolipoprotein E, apolipoprotein E3, late onset), MGC1571
Host Species Mouse
Immunogen Purified human apolipoprotein E/ApoE
Purification Method Protein G purified
Quantity 1000 μg
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cancer, Cholesterol Metabolism, Cognition and Behavior, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 348
Target Species Non-human Primate
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.