Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein E R2/ApoE R2 Antibody, Novus Biologicals™
SDP

Catalog No. NB1002216SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002216SS 0.025 mL
NB1002216 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1002216SS Supplier Novus Biologicals Supplier No. NB1002216SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

Apolipoprotein E R2/ApoE R2 Polyclonal specifically detects Apolipoprotein E R2/ApoE R2 in Chicken, Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, SDS-Page, Western Blot.

Specifications

Antigen Apolipoprotein E R2/ApoE R2
Applications Immunocytochemistry, Immunofluorescence, SDS-Page, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:200-1:500, SDS-Page reported in scientific literature (PMID 27810638)
Formulation Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
Gene Accession No. Q14114
Gene Alias APOER2HSZ75190, Apolipoprotein E receptor 2, low density lipoprotein receptor-related protein 8, apolipoprotein e receptor, low-density lipoprotein receptor-related protein 8, LRP-8, MCI1
Gene Symbols LRP8
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human ApoER2 protein sequence (between residues 863-963). [UniProt# Q14114]
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7804
Test Specificity Human, chicken and mouse. Immunogen has 95% homology to bovine.
Target Species Chicken, Mouse, Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.