Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein E R2/ApoE R2 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N102217A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N102217A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N102217A350 Supplier Novus Biologicals Supplier No. NB1002217AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Apolipoprotein E R2/ApoE R2 Polyclonal specifically detects Apolipoprotein E R2/ApoE R2 in Bovine, Chicken, Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen Apolipoprotein E R2/ApoE R2
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias APOER2HSZ75190, Apolipoprotein E receptor 2, low density lipoprotein receptor-related protein 8, apolipoprotein e receptor, low-density lipoprotein receptor-related protein 8, LRP-8, MCI1
Gene Symbols LRP8
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human ApoER2 protein sequence (between residues 800-900). [UniProtNo. Q14114]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7804.0
Target Species Bovine, Chicken, Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.