Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein E R2/ApoE R2 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB1002216Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002216Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002216Y Supplier Novus Biologicals Supplier No. NB1002216AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Apolipoprotein E R2/ApoE R2 Polyclonal specifically detects Apolipoprotein E R2/ApoE R2 in Chicken, Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, SDS-Page, Western Blot.

Specifications

Antigen Apolipoprotein E R2/ApoE R2
Applications Immunocytochemistry, Immunofluorescence, SDS-Page
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q14114
Gene Alias APOER2HSZ75190, Apolipoprotein E receptor 2, low density lipoprotein receptor-related protein 8, apolipoprotein e receptor, low-density lipoprotein receptor-related protein 8, LRP-8, MCI1
Gene Symbols LRP8
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human ApoER2 protein sequence (between residues 863-963). [UniProt# Q14114]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7804
Test Specificity Human, chicken and mouse. Immunogen has 95% homology to bovine.
Target Species Chicken, Human, Chicken, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.