Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein E3/ApoE3 Antibody (960319), DyLight 755, Novus Biologicals™
SDP

Catalog No. MAB41442IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB41442IR 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB41442IR Supplier Novus Biologicals Supplier No. MAB41442IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Apolipoprotein E3/ApoE3 Monoclonal specifically detects Apolipoprotein E3/ApoE3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Apolipoprotein E3/ApoE3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 960319
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AD2, Apo-E, ApoE4, apolipoprotein E, LDLCQ5, LPG
Gene Symbols APOE
Host Species Mouse
Immunogen ApoE3 variant peptide found within amino acids 100-200.
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.