Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein H/ApoH Antibody (OTI4A11), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP270186G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270186G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270186G Supplier Novus Biologicals Supplier No. NBP270186G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Apolipoprotein H/ApoH Monoclonal specifically detects Apolipoprotein H/ApoH in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Apolipoprotein H/ApoH
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4A11
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Activated protein C-binding protein, Anticardiolipin cofactor, APC inhibitor, Apo-H, Apolipoprotein H, apolipoprotein H (beta-2-glycoprotein I), B2G1, B2GP1, B2GPI, Beta(2)GPI, beta-2-glycoprotein 1, Beta-2-glycoprotein I, BG
Gene Symbols APOH
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 120-345 of human Apolipoprotein H/ApoH (NP_000033) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cholesterol Metabolism, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 350.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.