Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein L1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP106017 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106017 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106017 Supplier Novus Biologicals Supplier No. NBP106017
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Apolipoprotein L1 Polyclonal specifically detects Apolipoprotein L1 in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen Apolipoprotein L1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. O14791
Gene Alias ApoL, APO-L, ApoL-I, Apolipoprotein L, apolipoprotein L, 1, apolipoprotein L1, Apolipoprotein L-I, FSGS4
Gene Symbols APOL1
Host Species Goat
Immunogen Peptide with sequence C-NNNYKILQADQE corresponding to C-Terminus according to NP_003652.2, NP_663318.1.
Molecular Weight of Antigen 44 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8542
Test Specificity This antibody is expected to recognize both reported isoforms (NP_003652.2; NP_663318.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.