Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein L1 Rabbit anti-Human, Mouse, Clone: 1A8N2, Novus Biologicals™
SDP

Catalog No. NB167906 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167906 20 μg
NB167905 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167906 Supplier Novus Biologicals Supplier No. NBP31640720UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Apolipoprotein L1 Monoclonal antibody specifically detects Apolipoprotein L1 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen Apolipoprotein L1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 1A8N2
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ApoL, APO-L, ApoL-I, Apolipoprotein L, apolipoprotein L, 1, apolipoprotein L1, Apolipoprotein L-I, FSGS4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 303-398 of human Apolipoprotein L1 (O14791).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 8542
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.