Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APP Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB133530 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB133530 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB133530 Supplier Novus Biologicals Supplier No. NBP306205V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

APP Polyclonal specifically detects APP in Human samples. It is validated for ELISA.

Specifications

Antigen APP
Applications ELISA
Classification Polyclonal
Conjugate DyLight 405
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human APP (P05067-1; Asp672-Val711).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 351
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.