Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APPBP1 Antibody (OTI1E10), HRP, Novus Biologicals™
SDP

Catalog No. NBP272930H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272930H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272930H Supplier Novus Biologicals Supplier No. NBP272930H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

APPBP1 Monoclonal specifically detects APPBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NEDD8 Activating Enzyme (APPBP1/UBA3)
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1E10
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias A-116A10.1, amyloid beta precursor protein binding protein 1, amyloid beta precursor protein binding protein 1, 59kDa, Amyloid beta precursor protein-binding protein 1, 59 kDa, amyloid beta precursor protein-binding protein 1, 59kD, Amyloid protein-binding protein 1, APPBP1APP-BP1, NEDD8 activating enzyme E1 subunit 1, NEDD8-activating enzyme E1 regulatory subunit, NEDD8-activating enzyme E1 subunit, protooncogene protein 1, Proto-oncogene protein 1, ula-1
Gene Symbols NAE1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-274 of human NEDD8 Activating Enzyme (APPBP1/UBA3) (NP_001018170) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8883.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.