Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APPD Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179945
Description
APPD Polyclonal specifically detects APPD in Human samples. It is validated for Western Blot.Specifications
APPD | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
Apoptosis-inducing protein, APPDapoptosis-inducing protein D, LAPF, Lysosome-associated apoptosis-inducing protein containing PH and FYVE domains, MGC4090, PH and FYVE domain-containing protein 1, PH domain-containing family F member 1, phafin-1, PHAFIN1, pleckstrin homology domain containing, family F (with FYVE domain) member 1, pleckstrin homology domain-containing family F member 1, ZFYVE15phafin 1, Zinc finger FYVE domain-containing protein 15 | |
Rabbit | |
31 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%Yeast: 100%; Trypanosoma brucei: 100%; Trypanosoma brucei gambiense DAL972: 100%; Bovine: 92%; Canine: 92%; Mouse: 92%; Rat: 92%; Roseovarius sp. TM1035: 91%; Candida lipolytica: 90%; Brown alga: 90%; Phytophthora infestans T30-4: 90%; Erythrobacter sp. SD-21: 90%; Aspergillus clavatus: 83%; Caenorhabditis vulgaris: 83%; Xenopus: 83%; Fruit fly: 76%. | |
Human, Mouse, Rat, Bovine, Canine, Yeast | |
Purified |
Western Blot | |
1 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_077286 | |
PLEKHF1 | |
Synthetic peptide directed towards the C terminal of human APPDThe immunogen for this antibody is APPD. Peptide sequence QPAHLARPICGASSGDDDDSDEDKEGSRDGDWPSSVEFYASGVAWSAFHS. | |
Protein A purified | |
RUO | |
79156 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction