Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APPD Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | APPD |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
APPD Polyclonal specifically detects APPD in Human samples. It is validated for Western Blot.Specifications
APPD | |
Polyclonal | |
Purified | |
RUO | |
Apoptosis-inducing protein, APPDapoptosis-inducing protein D, LAPF, Lysosome-associated apoptosis-inducing protein containing PH and FYVE domains, MGC4090, PH and FYVE domain-containing protein 1, PH domain-containing family F member 1, phafin-1, PHAFIN1, pleckstrin homology domain containing, family F (with FYVE domain) member 1, pleckstrin homology domain-containing family F member 1, ZFYVE15phafin 1, Zinc finger FYVE domain-containing protein 15 | |
PLEKHF1 | |
IgG | |
Protein A purified |
Western Blot | |
Unconjugated | |
Rabbit | |
NP_077286 | |
79156 | |
Synthetic peptide directed towards the C terminal of human APPDThe immunogen for this antibody is APPD. Peptide sequence QPAHLARPICGASSGDDDDSDEDKEGSRDGDWPSSVEFYASGVAWSAFHS. | |
Primary | |
31 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title