Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APPL Rabbit anti-Human, Mouse, Rat, Clone: 4M5X9, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31672120UL
This item is not returnable.
View return policy
Description
APPL Monoclonal antibody specifically detects APPL in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
APPL | |
Monoclonal | |
Unconjugated | |
Adapter protein containing PH domain, PTB domain and leucine zipper motif 1, adaptor protein containing pH domain, PTB domain and leucine zipper motif 1, adaptor protein, phosphotyrosine interaction, PH domain and leucine zippercontaining 1, AKT2 interactor, APPLdip13-alpha, DCC-interacting protein 13-alpha, DIP 13 alpha, DIP13A, Dip13-alpha, DIP13alpha, KIAA1428, signaling adaptor protein DIP13alpha | |
A synthetic peptide corresponding to a sequence within amino acids 350-450 of human APPL1 (Q9UKG1). | |
20 μg | |
Apoptosis, Cell Biology, Neuroscience, Signal Transduction | |
26060 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
4M5X9 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction