Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APPL Rabbit anti-Human, Mouse, Rat, Clone: 4M5X9, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | APPL |
---|---|
Clone | 4M5X9 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
APPL Monoclonal antibody specifically detects APPL in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
APPL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cell Biology, Neuroscience, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
26060 | |
IgG | |
Affinity purified |
4M5X9 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Adapter protein containing PH domain, PTB domain and leucine zipper motif 1, adaptor protein containing pH domain, PTB domain and leucine zipper motif 1, adaptor protein, phosphotyrosine interaction, PH domain and leucine zippercontaining 1, AKT2 interactor, APPLdip13-alpha, DCC-interacting protein 13-alpha, DIP 13 alpha, DIP13A, Dip13-alpha, DIP13alpha, KIAA1428, signaling adaptor protein DIP13alpha | |
A synthetic peptide corresponding to a sequence within amino acids 350-450 of human APPL1 (Q9UKG1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title