Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ AmpliTaq Gold™ DNA Polymerase, LD (Low DNA) with Gold Buffer and MgCl2

Catalog No. p-4919370
Encompass_Preferred
Click to view available options
Quantity:
1000 Units
250 Units

Catalog No. 43-388-56

Supplier: Applied Biosystems™ 4338856

Only null left
Add to Cart
Add to Cart

Includes

Contains AmpliTaq Gold DNA Polymerase, LD (250 units), GeneAmp 10X PCR Gold Buffer, and MgCl2 Solution.

Applied Biosystems AmpliTaq Gold DNA Polymerase, LD (Low DNA) is a highly purified version of AmpliTaq Gold DNA Polymerase that significantly reduces contaminating bacterial DNA sequences commonly present in recombinant protein preparations.

Applied Biosystems AmpliTaq Gold DNA Polymerase, LD (Low DNA), is a highly purified version of AmpliTaq Gold DNA Polymerase that significantly reduces contaminating bacterial DNA sequences commonly present in recombinant protein preparations.

This highly purified enzyme preparation gives you more confidence in your results and makes advanced PCR techniques quick and easy. When combined with the supplied GeneAmp 10X PCR Gold Buffer, AmpliTaq Gold DNA Polymerase, LD, provides a flexible hot-start system that is suitable for all PCR applications that require the elimination of bacterial DNA contamination.

Features

  • Highly purified enzyme with lower levels of bacterial DNA
  • Minimized non-target, false-positive PCR products when amplifying bacterial sequences
  • Chemically modified hot-start technique eliminates biological contamination
  • 10X PCR Gold Buffer promotes fast enzyme activation and robust PCR amplification
  • Enhanced specificity with time-release PCR for low-copy target amplification
  • Improved yield and success rate for target amplification

Notes

  • AmpliTaq Gold DNA Polymerase, LD, is recommended for PCR applications that require low background levels of bacterial DNA.
  • See user's manual or package insert for limited label license and trademark information.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Content And Storage • AmpliTaq Gold DNA Polymerase, LD (5 U/μL), 1 x 50 μL
• 10X PCR Gold Buffer, 1 x 1.5 mL
• 25 mM MgCl2, 1 x 1.5 mL

Store at -15°C to -30°C.
Format Tube
GC-Rich PCR Performance Low
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
Exonuclease Activity 5' - 3'
Product Type DNA Polymerase
Purity or Quality Grade LD (Low DNA)
Quantity 250 Units
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR
Fidelity (vs. Taq) 1X
Hot Start Built-In Hot Start
No. of Reactions 200 Reactions
Overhang 3'-A
Reaction Format Separate Components
Size (Final Product) 5 kb or less
Starting Material DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.